Learn More
Beckman Coulter CD8, Single Color Antibody, IM0451U
Supplier:  Beckman Coulter IM0451U
Encompass
The Beckman Coulter CD8 Antibody (IM0451U) is a purified mouse monoclonal antibody that binds to human CD8 antigen. It is intended for use in flow cytometry to identify cytotoxic/suppressor T-lymphocytes. This antibody is supplied at 0.5 mg/mL in phosphate-buffered saline with 0.1% sodium azide, and has a 12 month shelf life when stored at 2-8°C. The recommended working dilution is 5-20 μL per test, with titration recommended for optimal staining. This antibody is useful for immunophenotyping studies.
Catalog No. NC1832987