Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cd80-in-3 | 486449-65-4 | MFCD32701897 | 98.0% | 323.28 g/mol | C17H10FN3O3 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10089150MG

Encompass_Preferred

CD80-IN-3 is a small-molecule inhibitor that blocks the CD80-CD28 interaction, with reported EC50 of 630 nM and a Kd of 125 nM. The compound is intended for research use only and is supplied as a solid or as DMSO solutions.

  • Potent CD80 inhibitor (EC50 630 nM; Kd 125 nM).
  • High purity, 98.02% (by HPLC).
  • Molecular weight 323.28 g/mol and formula C17H10FN3O3.
  • Soluble in DMSO (35.71 mg/mL); may require ultrasonic treatment.
  • Available as solid and as 10 mM solutions in DMSO.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (2 years) or -20 °C (1 year).
  • Light brown to yellow solid appearance.

Catalog No. 50-002-13093


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.