Learn More
Cusabio Technology LLC Human CD81 antigen(CD81) ELISA kit
Supplier:  Cusabio Technology LLC CSBEL004960HU
Encompass_Preferred
This Human CD81 ELISA Kit was designed for the quantitative measurement of Human CD81 protein in serum, plasma, tissue homogenates, cell lysates. It is a Sandwich ELISA kit, its detection range is 0.156 ng/mL-10 ng/mL and the sensitivity is 0.039 ng/mL. One kit contains 96 wells.
Catalog No. NC2952977