Learn More
FUJIFILM BIOSCIENCES INC CD9-Capture Human Exosome ELIS
Supplier:  FUJIFILM BIOSCIENCES INC 29683701
Encompass_Preferred
The CD9-Capture Human Exosome ELISA Kit Streptavidin HRP utilizes an anti-human CD9 antibody pre-coated onto a 96-well plate for selective capture of CD9-positive extracellular vesicles The use of biotinylated detection antibodies and HRP-conjugated streptavidin enables high-sensitivity detection via TMB substrate The kit is compatible with a wide range of biological samples including serum plasma and culture supernatants Its versatility allows use with additional biotinylated antibodies for multiplexed exosome profiling making it a powerful tool in exosome research and biomarker discovery
Catalog No. 50-004-50331