Learn More
Medchemexpress LLC CD96 Human HEK293 10ug
SDP
Supplier:  Medchemexpress LLC HYP7269810UG
Encompass_Preferred
CD96 promotes adhesive interactions between activated T cells and NK cells in the late stages of the immune response It promotes the adhesion between NK cells and target cells by interacting with PVR on target cells CD96 Protein Human (HEK293 His) is the recombinant human-derived CD96 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82250