Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD96 / TACTILE Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;TAE,CD96,Tactile
Supplier:  ACROBiosystems TAEH82E3200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD96 / TACTILE Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human CD96, His Tag, Avi Tag (TAE-H82E3) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Met 503 (Accession # P40200-2).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 57.3 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CD96, His Tag, Avi Tag (Cat. No. TAE-H82E3) at 5 μg/mL (100 μL/well) can bind Human CD155, Fc Tag (Cat. No. CD5-H5251) with a linear range of 39-625 ng/mL (QC tested).
Catalog No. 50-165-7940