Learn More
Medchemexpress LLC CD97 protein, human (HEK293, hFc) | >90.0% | 50 UG

Supplier: Medchemexpress LLC HYP7537050UG
Recombinant human CD97 Fc-tagged protein (residues Q21-Q398 fused to C-terminal human Fc) expressed in HEK293 mammalian cells. Supplied as a sterile, 0.22 μm-filtered PBS solution (pH 7.4), the glycosylated protein is intended for biochemical and binding assays and demonstrates binding to human CD55 in ELISA.
- Recombinant fragment Q21-Q398 fused to human Fc.
- Expressed in HEK293 cells with mammalian glycosylation.
- Purity greater than 90% by reducing SDS-PAGE.
- Endotoxin level below 1 EU/μg (LAL method).
- Molecular weight approximately 100-110 kDa.
- Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
- Measured binding to CD55 with ELISA (linear range 1.28-32 ng/mL).
- Shipped on dry ice; store at -80°C and avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.