Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cdc7-IN-5 | 1402057-86-6 | 96.7% | 445.47 g/mol | C25H23N3O5 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1305175MG

Encompass_Preferred

Cdc7-IN-5 is a small-molecule inhibitor of the Cdc7 serine-threonine kinase supplied for research use. The compound is characterized by molecular formula C25H23N3O5, molecular weight 445.47 g/mol, and reported purity of about 96.7%. It is provided as a solid powder with documented solubility in DMSO and recommended storage conditions for powder and solutions.

  • Potent inhibitor of Cdc7 kinase for cell cycle and DNA replication studies.
  • Molecular formula C25H23N3O5; molecular weight 445.47 g/mol.
  • High purity (about 96.7%).
  • Soluble in DMSO; warming and sonication recommended to aid dissolution.
  • Powder and solution storage conditions specified for long-term stability.
  • Includes datasheet, SDS, COA, and analytical data for quality verification.

Catalog No. 50-002-62110


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.