Learn More
Medchemexpress LLC CDDO-2P-Im | 1883650-96-1 | 98.1% | 618.81 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12637910MG
Encompass_Preferred
CDDO-2P-Im is an analogue of CDDO-Imidazolide with a chemopreventive effect. It can reduce the size and severity of lung tumors in a mouse lung cancer model and is more stable than CDDO-Im in pharmacokinetic studies. It induces differentiation of U937 cells and suppresses NO production in RAW264.7 cells.
  • Induces differentiation of U937 cells at 100 nM
  • Suppresses NO production in RAW264.7 cells
  • Elevates heme oxygenase-1 (HO-1) and quinone reductase (NQO1) mRNA and protein levels
  • Decreases the number, size, and severity of tumors in A/J mice
Catalog No. 50-004-36394