Learn More
Selleck Chemical LLC CDDO-Im S9723-25MG
SDP
Supplier:  Selleck Chemical LLC S972325MG
Encompass_Preferred
CDDO-Im (RTA-403) is an activator of nuclear factor erythroid 2 related factor 2 (Nrf2) and peroxisome proliferator-activated receptor (PPAR) CDDO-Im binds to PPAR and PPAR with Ki of 232 nM and 344 nM respectively CDDO-Im inhibits inflammatory response and tumor growth in vivo
Catalog No. 50-000-24416