Learn More
Bioss Cdk1 - Tyr-15 conserved site
SDP
Supplier:  Bioss BSM70419M
Encompass_Preferred
Cdk1 (Tyr-15)[conserved site] Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Clonality Monoclonal Applications WB M231Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65793