Learn More
Sino Biological Recombinant Mouse CCRK Protein (Full Length, GST Tag)
Supplier:  Sino Biological C2130G1000
Encompass_Preferred
Recombinant full length mouse CDK20 (CCRK) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5656