Learn More
Medchemexpress LLC CDK12-IN-2 | 2244987-03-7 | 99.4% | 532.64 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112626100MG
Encompass_Preferred
CDK12-IN-2 is a potent, selective, and nanomolar CDK12 inhibitor (IC50=52 nM) with good physicochemical properties. It is also a strong CDK13 inhibitor, as CDK13 is the closest homologue of CDK12. It inhibits the phosphorylation of Ser2 in the C-terminal domain of RNA polymerase II and can be used as an excellent chemical probe for functional studies of CDK12.
  • Potent, selective, and nanomolar CDK12 inhibitor
  • Strong CDK13 inhibitor
  • Excellent kinase selectivity for CDK12 over CDK2, 9, 8, and 7
  • Inhibits phosphorylation of Ser2 in the C-terminal domain of RNA polymerase II
  • Useful as a chemical probe for functional studies of CDK12
Catalog No. 50-004-36190