Learn More
Medchemexpress LLC CDK12-IN-2 | 2244987-03-7 | 99.4% | 532.64 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11262650MG
Encompass_Preferred
CDK12-IN-2 is a potent, selective, and nanomolar CDK12 inhibitor with good physicochemical properties. It also acts as a strong CDK13 inhibitor and exhibits excellent kinase selectivity over CDK2, 9, 8, and 7. This compound inhibits the phosphorylation of Ser2 in the C-terminal domain of RNA polymerase II and can be used as a chemical probe for functional studies of CDK12.
  • Potent, selective, and nanomolar CDK12 inhibitor (IC50=52 nM)
  • Strong CDK13 inhibitor
  • Excellent kinase selectivity for CDK12 over CDK2, 9, 8, and 7
  • Inhibits phosphorylation of Ser2 in the C-terminal domain of RNA polymerase II
  • Can be used as a chemical probe for functional studies of CDK12
  • Inhibits CTD Ser2 phosphorylation in SK-BR-3 cells (IC50 of 185 nM)
  • Exhibits growth inhibition in SK-BR-3 cells (IC50 of 0.8 μM)
Catalog No. 50-004-37466