Learn More
Medchemexpress LLC CDK12-IN-E9 5mg | 2020052-55-3 | 434.53 | C24H30N6O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY117203A5MG
Encompass_Preferred
CDK12-IN-E9 is a research-stage kinase inhibitor used to probe CDK12-mediated transcriptional regulation, DNA damage response, and cell-cycle processes. It is supplied as a high-purity solid and as a 10 mM solution in DMSO, and is intended for laboratory research use only.
  • Potent, selective covalent CDK12 inhibition.
  • Non-covalent inhibitory activity against CDK9.
  • High reported purity (99.71%).
  • Available as solid and DMSO solution formats.
  • Suitable for biochemical and cell-based research applications.
Catalog No. 50-002-26501