Learn More
Cayman Chemical Cdk15 InhIbItr 10mg
SDP
Supplier:  Cayman Chemical 1874010 MG
Encompass_Preferred
Inhibitor of Cdk1/cyclin B and Cdk/p25 (IC50s = 600 and 400 nM, respectively)
Catalog No. 50-296-7805