Learn More
Medchemexpress LLC HY-117535 10mg Medchemexpress, CDK2-IN-4 CAS:2079895-42-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11753510MG
Encompass_Preferred
Medchemexpress, HY-117535 10mg CDK2-IN-4 CAS:2079895-42-2 CDK2-IN-4 is a potent and selective CDK2 inhibitor with an IC50 of 44 nM for CDK2/cyclin A, shows 2,000-fold selectivity over CDK1/cyclin B (IC50=86 uM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9477