Learn More
Medchemexpress LLC Cdk2-in-4 | 2079895-42-2 | MFCD32174273 | 99.9% | 442.49 g/mol | C23H18N6O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11753510MM 1ML
Encompass_Preferred
CDK2-IN-4 is a potent, selective small-molecule inhibitor of cyclin-dependent kinase 2 (CDK2). It inhibits CDK2/cyclin A with an IC50 of 44 nM and shows approximately 2,000-fold selectivity over CDK1/cyclin B (IC50 = 86 μM). The compound is intended for research use in cell-cycle and cancer biology studies.
  • Potent CDK2 inhibition (IC50 = 44 nM).
  • High selectivity versus CDK1 (≈2,000-fold; CDK1 IC50 = 86 μM).
  • Supplied as a 10 mM solution in DMSO (1 mL) and as solid material in multiple package sizes.
  • Molecular weight 442.49 g/mol; chemical formula C23H18N6O2S.
  • Datasheet and safety data sheet available for laboratory handling and safety information.
Catalog No. 50-002-27673