Learn More
Selleck Chemical LLC CDK2-IN-73 S0273-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S027310MM/1ML
Encompass_Preferred
CDK2-IN-73 (CDK2-IN-4 CDK2 inhibitor 73) is a potent and selective inhibitor of CDK2 with IC50 of 44 nM for CDK2/cyclin A
Catalog No. 50-000-43581