Learn More
Cayman Chemical Cdk2 InhIbItr II 500ug
SDP
Supplier:  Cayman Chemical 15154500 UG
Encompass_Preferred
A 3-(benzylidne)indolin-2-one analog that selectively and potently inhibits Cdk2 (IC50 = 60 nM)
Catalog No. 50-297-2778