Learn More
Sino Biological Recombinant Human CDK2 & Cyclin E2 Protein (Full Length, GST Tag), Active
Supplier:  Sino Biological C2918BG10
Encompass_Preferred
Recombinant full length human CDK2 and CyclinE2 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
Catalog No. 50-256-9576