Learn More
Cayman Chemical Cdk4 InhIbItr 5mg
SDP
Supplier:  Cayman Chemical 176485 MG
Encompass_Preferred
A cell-permeable, asymmetrical indolocarbazole that exhibits antiproliferative activity by selectively blocking cyclin D1/Cdk4 (IC50 = 0.8 μM); inhibits HCT116 and NCI-H460 tumor cell growth (IC50s 1 cell cycle arrest
Catalog No. 50-296-7060