Learn More
Cayman Chemical Cdk46 InhIbItr IV 10mg
SDP
Supplier:  Cayman Chemical 1797410 MG
Encompass_Preferred
A cell-permeable triaminopyrimidine that selectively and reversibly blocks Cdk4/cyclin D1 and Cdk6/cyclin D1 activity (IC50s = 1.5 and 5.6 μM, respectively); induces cell cycle arrest in the G1 phase and apoptosis in asynchronous cell lines; suppresses tumor growth in mice bearing human HCT116 colon carcinoma xenografts
Catalog No. 50-296-7573