Learn More
Medchemexpress LLC CDK5 inhibitor 20-223 | 865317-30-2 | 99.6% | 305.37 g·mol⁻¹ | C19H19N3O | 10MG

Supplier: Medchemexpress LLC HY12377210MG
CDK5 inhibitor 20-223 is a potent small-molecule inhibitor of cyclin-dependent kinases CDK2 and CDK5, reported to inhibit these kinases with IC50s of 6.0 nM and 8.8 nM, respectively. It has demonstrated antiproliferative activity in colorectal cancer cell lines and antitumor efficacy in xenograft models. The compound is supplied as a light yellow solid with high purity and recommended storage conditions for powder and solutions.
- Potent CDK2 and CDK5 inhibitor with reported IC50s of 6.0 nM and 8.8 nM.
- Demonstrated antiproliferative activity in colorectal cancer cell lines and in vivo efficacy.
- Molecular formula C19H19N3O and molecular weight 305.37 g·mol⁻¹.
- High purity (≈99.6%) and supplied as a light yellow solid.
- Recommended storage: powder at -20°C (long term) or 4°C; solutions at -80°C or -20°C.
- Intended for biochemical and cell-based research into CDK biology.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.