Learn More
ABclonal Technology CDK5RAP2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A154761000UL
Encompass_Preferred
This gene encodes a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein encoded by this gene is localized to the centrosome and Golgi complex, interacts with CDK5R1 and pericentrin (PCNT), plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimers disease. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-06277