Learn More
Medchemexpress LLC Cdk9-in-10 | 3542-63-0 | 98.1% | C22H16O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13085010MG
Encompass_Preferred
CDK9-IN-10 is a small-molecule research reagent that acts as a potent inhibitor of cyclin-dependent kinase 9 (CDK9) and can serve as the ligand component in PROTAC degrader design. It is supplied as a high-purity compound for laboratory biochemical and cellular studies and is intended for research use only.
  • Potent inhibitor of cyclin-dependent kinase 9 (CDK9).
  • High reported purity suitable for research applications.
  • Useful as a ligand in targeted protein degradation studies.
  • Available in multiple small-molecule package sizes for assay development.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-59883