Learn More
Medchemexpress LLC Cdk9-in-30 | 748146-89-6 | 98.0% | C16H20FNO3 | 10MG

Supplier: Medchemexpress LLC HY14713110MG
CDK9-IN-30 is a small-molecule research inhibitor of cyclin-dependent kinase 9 (CDK9) supplied as a purified compound for in vitro and preclinical studies. It has reported antiviral activity, including inhibition of HIV-1 replication, and is typically prepared in polar aprotic solvents for assay use.
- Inhibits cyclin-dependent kinase 9 in biochemical and cellular assays.
- Shows reported inhibition of HIV-1 replication in published studies.
- High purity suitable for research applications.
- Molecular formula C16H20FNO3 and molecular weight 293.33 g·mol⁻¹.
- Soluble in DMSO; prepare stock solutions in DMSO for assays.
- Packaged in small research quantities and stored refrigerated to maintain stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.