Learn More
TARGETMOL CHEMICALS INC CDK9-IN-7 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T107455MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CDK9-IN-7 is a highly selective and orally active CDK9/cyclin T inhibitor (IC50 11 nM) which exhibits more potent over other CDKs (CDK4/cyclinD 148 nM; CDK6/cyclinD 145 nM). purity: 98%
Catalog No. 50-258-1675