Learn More
Medchemexpress LLC HY-102039 5mg Medchemexpress, CDK9-IN-8 CAS:2105956-51-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1020395MG
Encompass_Preferred
Medchemexpress, HY-102039 5mg CDK9-IN-8 CAS:2105956-51-0 CDK9-IN-8 is a highly effective and selective CDK9 inhibitor with an IC50 of 12 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7966