Learn More
Medchemexpress LLC Cdk9 inhibitor HH1 | 204188-41-0 | 99.7% | 261.34 g/mol | C13H15N3OS | 5MG

Supplier: Medchemexpress LLC HY1246395MG
CDK9 inhibitor HH1 is a potent, selective small-molecule inhibitor of cyclin-dependent kinase 9 (CDK9) used in biochemical and cellular research to study transcriptional regulation and cancer biology.
- Potent and selective CDK9 inhibition.
- Inhibits CDK9-mediated transcription.
- Suitable for cancer biology and transcription research.
- Molecular formula C13H15N3OS; molecular weight 261.34 g·mol⁻1.
- Purity approximately 99.7% as reported.
- Appearance: light yellow to yellow solid.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Available pack sizes: 5 mg, 10 mg, 50 mg, 100 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.