Learn More
Sino Biological Recombinant Human CDK7 & CCNH & MNAT1 Protein (Full Length, His Tag), Active
Supplier:  Sino Biological C3610H500
Encompass_Preferred
Recombinant full length human CDK7, Cyclin H1 and MNATI were co-expressed by baculovirus in Sf9 insect cells using N-terminal His tags.
Catalog No. 50-257-5554