Learn More
Medchemexpress LLC HY-12445 10mg , CDKI-73 CAS:1421693-22-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1244510MG
Encompass_Preferred
HY-12445 10mg CDKI-73 CAS: 1421693-22-2 CDKI-73 (LS-007) is an orally active and highly efficacious CDK9 inhibitor, with Ki values of 4 nM, 4 nM and 3 nM for CDK9, CDK1 and CDK2, respectively. CDKI-73 down-regulates the RNAPII phosphorylation. CDKI-73 is also a novel pharmacological inhibitor of Rab11 cargo delivery and innate immune secretion[4].
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9995