Learn More
Genetex Inc CDKN2A / p16INK4a Monoclonal Antibody [MSVA-016R] HistoMAX™ (GTX04442) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04442
Encompass_Preferred
The CDKN2A / p16INK4a antibody [MSVA-016R] HistoMAX™ (GTX04442) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human squamous cell carcinoma (SCC) from uterine tissue using GTX04442 CDKN2A / p16INK4a antibody [MSVA-016R] HistoMAX. Squamous cell carcinoma of the cervix uteri HPV positive with strong CDKN2A / p16INK4a immunostaining.
Catalog No. NC3666959