Learn More
Medchemexpress LLC CDKN2A/p16INK4a antibody (YA508)(PBS only) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP80615A10UL
Encompass_Preferred
CDKN2A/p16INK4a antibody (YA508) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets CDKN2A/p16INK4a.
  • Reacts with human and mouse samples
  • Suitable for western blot (WB) applications
  • Suitable for immunoprecipitation (IP) applications
Catalog No. 50-003-96130