Learn More
Lonza Walkersville CDM Chondrocyte Differentiation Medium BulletKit
Supplier:  Lonza Walkersville CC3225
Encompass
CDM Chondrocyte Differentiation Medium is specially designed for the differentiation of chondrocytes. The BulletKit is conveniently packaged with basal medium and supplements included for the differentiation of chondrocytes.CGM Chondrocyte Growth Medium is also available and sold separately for expansion of human chondrocytes.Content:1 x Bottle Basal Medium 250 mL1 x Yellow Cap Vial with R3-IGF 0.50 mL1 x Blue Cap Vial with TGF-beta 1.25 mL1 x Lilac Cap Vial with Insulin 0.50 mL1 x Red Cap Vial with GA-1000 0.25 mL1 Bottle FBS 12.50 mL1 x Natural Cap Vial with Transferrin 0.50 mL
Catalog No. NC9054301
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.