Learn More
Biochain Institute Inc cDNA - Dog Normal Tissue: Stomach, 40 rxn/PK
SDP
Supplier:  Biochain Institute Inc C1734248
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different dog tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-846