Learn More
Biochain Institute Inc cDNA - Human Diabetic Diseased Tissue: Small Intestine: Duodenum, 40 rxn/PK
SDP
Supplier:  Biochain Institute Inc C1236101DIA
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human diabetic diseased tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-179-979