Learn More
Biochain Institute Inc cDNA - Lupus: Liver, 40 rxn/PK
SDP
Supplier:  Biochain Institute Inc C1236149LUP
Encompass_Preferred
PCR Ready First Strand cDNAs from human lupus diseased tissues are available. More organs maybe available upon request. All the cDNAs are for research use only. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-090