Learn More
Biochain Institute Inc cDNA - Human Tumor Cell Line: Hela, 40 rxn/PK
SDP
Supplier:  Biochain Institute Inc C1255811
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from tumor cell lines. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-209