Learn More
Biochain Institute Inc cDNA - Monkey (Cynomolgus) Normal Tissue: Spinal Cord, 40 rxn/PK
SDP
Supplier:  Biochain Institute Inc C1534234CY
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from different monkey normal tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-268