Learn More
Biochain Institute Inc cDNA - Parkinson's Disease: Brain: Hippocampus, 40 rxn/PK
SDP
Supplier:  Biochain Institute Inc C1236052PAR
Encompass_Preferred
BioChain's PCR Ready First Strand cDNAs are available from human neurological diseased brain tissues. 1 ul cDNA is sufficient for each PCR reaction.
Catalog No. 50-180-125