Learn More
Hach Company CDTA Reagent, 30 mL SCDB
Supplier:  Hach Company 2589723
Encompass
For ultra-low hardness determination by the Chlorophosphonazo method (Hach method 8374). 30 mL Self-Contained Dropping Bottle.
Catalog No. 50-199-6285