Learn More
Medchemexpress LLC CE-224535 (PF-04905428) | 724424-43-5 | C22H29ClN4O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1548750MG
Encompass_Preferred
CE-224535 is a selective P2X7 receptor antagonist developed as a disease-modifying antirheumatic drug (DMARD). It acts as a selective antagonist of the human P2X7 receptor, reducing the secretion of IL-1 and IL-18 from leukocytes, suggesting a potential new therapeutic approach for the treatment of rheumatoid arthritis (RA).
  • Selective P2X7 receptor antagonist
  • Developed as a disease-modifying antirheumatic drug (DMARD)
  • Reduces leukocyte secretion of IL-1 and IL-18
  • Potential therapeutic approach for rheumatoid arthritis
  • For research use only
Catalog No. 50-003-76611