Learn More
Medchemexpress LLC CE-245677 | 717899-97-3 | 98.1% | 513.38 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112423100MG
Encompass_Preferred
CE-245677 is a potent reversible inhibitor of Tie2 and TrkA/B kinases with cellular IC50s of 4.7 and 1 nM. It also demonstrates over 100x selectivity against other angiogenic receptor tyrosine kinases, such as KDR, PDGFR, and FGFR. It shows good oral absorption in in vivo rat PK studies with a bioavailability of 80%.
  • Potent reversible inhibitor of Tie2 and TrkA/B kinases.
  • Cellular IC50s of 4.7 nM for Tie2 and 1 nM for TrkA/B.
  • More than 100x selectivity against other angiogenic receptor tyrosine kinases (KDR, PDGFR, FGFR).
  • Good oral absorption in in vivo rat PK studies (F=80%).
Catalog No. 50-004-35639