Learn More
Medchemexpress LLC CE-245677 | 717899-97-3 | 98.1% | 513.38 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11242350MG
Encompass_Preferred
CE-245677 is a potent reversible inhibitor of Tie2 and TrkA/B kinases with cellular IC50s of 4.7 nM and 1 nM, respectively. It exhibits >100x selectivity against other angiogenic receptor tyrosine kinases such as KDR, PDGFR, and FGFR.
  • Potent reversible inhibitor of Tie2 and TrkA/B kinases.
  • Cellular IC50s of 4.7 nM for Tie2 and 1 nM for TrkA/B.
  • High selectivity (>100x) against other angiogenic receptor tyrosine kinases (KDR, PDGFR, FGFR).
  • Shows good oral absorption in in vivo rat PK studies (F=80%).
Catalog No. 50-004-36729