Learn More
Medchemexpress LLC Ce-245677 | 717899-97-3 | 98.1% | 513.38 g/mol | C24H22Cl2N6O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1124235MG
Encompass_Preferred
CE-245677 is a potent, reversible small-molecule inhibitor of Tie2 and TrkA/B receptor tyrosine kinases, developed for preclinical research. It shows low-nanomolar cellular potency and has been characterized for in vitro assays and in vivo pharmacokinetic studies; the compound is provided as a powder with documented solubility and storage conditions to support experimental use.
  • Potent reversible inhibitor of Tie2 and TrkA/B with low-nanomolar cellular activity.
  • High purity suitable for biochemical and cellular assays.
  • Powder form with defined cold-storage recommendations.
  • Readily soluble in DMSO; in vivo formulation guidance available.
  • Demonstrated oral bioavailability in rodent pharmacokinetic studies.
Catalog No. 50-002-63526