Learn More
Genscript Corporation CEA RC66 mAb Mouse
Supplier:  Genscript Corporation V0620110
Encompass
Carcinoembroyonic antigen (CEA) is a glycoprotein produced during the embryonic and fetal period The high concentration of CEA in blood are observed in many patients with cancer of the lung liver pancreas breast colon and prostate The elevated CEA levels are related to the stage of the disease
Catalog No. 50-000-58118