Learn More
Medchemexpress LLC CEACAM1 Human N-Hi 5ug

Supplier: Medchemexpress LLC HYP72129A5UG
Recombinant human CEACAM1 protein with an N-terminal 6xHis tag, supplied as a lyophilized 5 μg vial for research use. The protein is produced in E. coli, provided at high purity, and formulated with Tris, NaCl, and arginine to improve solubility and stability for biochemical and binding assays.
- Recombinant human protein with N-terminal His tag for purification and detection.
- Expressed in E. coli and lyophilized for convenient storage and handling.
- High purity (>95%) confirmed by reducing SDS-PAGE.
- Approximate molecular weight ~45 kDa suitable for electrophoresis and western blot.
- Formulated to improve solubility and stability in aqueous buffers.
- Store at -20°C; aliquot to avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.