Learn More
Medchemexpress LLC Cearoin | 52811-37-7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN84181MG
Encompass_Preferred
Cearoin is a research chemical that promotes autophagy and apoptosis by stimulating ROS production and ERK activation. It is for research use only.
  • Increases autophagy
  • Increases apoptosis
  • Promotes ROS production
  • Activates ERK
  • Has a purity of 99.09%
  • Light yellow to green yellow solid appearance
Catalog No. 50-004-18725