Learn More
CPC Scientific H-Lys-Trp-Lys-Leu-Phe-Lys-Lys-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ATMP002A
Encompass
A 15-residue hybrid peptide incorporating partial sequences of cecropin A and melittin that causes the release of carboxyfluoresceine encapsulated in phosphatidylcholine liposomes.SEQUENCE: H-Lys-Trp-Lys-Leu-Phe-Lys-Lys-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: KWKLFKKIGAVLKVL-NH2MOLECULAR FORMULA: C89H152N22O15MOLECULAR WEIGHT: 1770.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [290814-90-3]SYNONYMS: Cecropin A (1-8)-Melittin A (3-9) amide, CAMEL0, Cecropin A (1-7)-Melittin A (2-9) amideRESEARCH AREA: AntimicrobialREFERENCES:D. Andreau et al., PNAS, 80, 6475 (1983)D. Andreau et al., Biochem., 24, 1683 (1985)SKU(s): ATMP-002A, ATMP-002B, ATMP-002C, ATMP-002D
Catalog No. 50-210-8808